Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231862.2 | XP_006231924.1 | 9 - 10 | 1641 | 78 | |
XM_006231863.2 | XP_006231925.1 | 9 - 10 | 1445 | 78 | ||
XM_008760695.1 | XP_008758917.1 | 13 - 14 | 1992 | 78 | ||
XM_008760696.1 | XP_008758918.1 | 12 - 13 | 1887 | 78 | ||
XM_008760697.1 | XP_008758919.1 | 12 - 13 | 1873 | 78 | ||
XM_008760698.1 | XP_008758920.1 | 8 - 9 | 1307 | 78 | ||
GenBank mRNA | AF361881.1 | - | 458 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |