Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013213.1 | NP_001013231.1 | 7 - 8 | 813 | 154 | |
XM_008763882.1 | XP_008762104.1 | 5 - 6 | 630 | 154 | ||
XM_008763883.1 | XP_008762105.1 | 8 - 9 | 555 | 154 | ||
XM_008763884.1 | XP_008762106.1 | 7 - 8 | 509 | 154 | ||
GenBank mRNA | BC086363.1 | - | 7 - 8 | 813 | 7304501 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |