Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114606.1 | NP_001108078.1 | 9 - 10 | 1631 | 103 | |
XM_006246509.2 | XP_006246571.1 | 9 - 10 | 3228 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |