Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009677.1 | NP_001009677.1 | 8 - 9 | 2749 | 88 | |
XM_006247527.2 | XP_006247589.1 | 8 - 9 | 2789 | 88 | ||
XM_008768292.1 | XP_008766514.1 | 7 - 8 | 2569 | 88 | ||
GenBank mRNA | BC087058.1 | - | 8 - 9 | 2749 | 7124252 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |