Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ214513.1 | - | 1 | 285 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |