Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008904.1 | NP_001008904.1 | 1 | 51 | 94 | |
GenBank mRNA | AY510277.1 | - | 1 | 51 | 94 |
Gene Symbol: LOC102551504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228159.1 | XP_006228221.1 | 1 | 51 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |