Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191642.1 | NP_001178571.1 | 3 - 4 | 2580 | 76 | |
XM_006246453.2 | XP_006246515.1 | 4 - 5 | 3022 | 76 | ||
XM_006246455.2 | XP_006246517.1 | 3 - 4 | 2683 | 76 | ||
XM_006246457.2 | XP_006246519.1 | 3 - 4 | 2683 | 76 | ||
XM_006246458.2 | XP_006246520.1 | 3 - 4 | 2685 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |