Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108598.2 | NP_001102068.2 | 4 - 5 | 388 | 90 | |
NM_001244784.1 | NP_001231713.1 | 4 - 5 | 388 | 90 | ||
GenBank mRNA | BC168211.1 | - | 4 - 5 | 217 | 7135378 | 90 |
CB576376.1 | - | 4 - 5 | 265 | 90 | ||
CB585518.1 | - | 4 - 5 | 388 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |