Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134338.1 | NP_599165.1 | 1 | 2122 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |