Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012854.2 | NP_036986.2 | 3 - 4 | 377 | 101 | |
XM_006249712.2 | XP_006249774.1 | 3 - 4 | 716 | 101 | ||
GenBank mRNA | L02926.1 | - | 3 - 4 | 377 | 101 | |
X60675.1 | - | 420 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Inventoried Probe spans exons |