Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244310.1 | NP_001231239.1 | 2 - 3 | 829 | 87 | |
GenBank mRNA | AY722391.1 | - | 1 - 2 | 393 | 87 | |
HQ657211.2 | - | 2 - 3 | 614 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |