Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244437.1 | NP_001231366.1 | 4 | 531 | 89 | |
XM_021068521.1 | XP_020924180.1 | 446 | 89 | |||
XM_021068522.1 | XP_020924181.1 | 273 | 89 | |||
GenBank mRNA | AK231928.1 | - | 4 | 531 | 89 | |
AK392765.1 | - | 4 | 487 | 89 | ||
AK393825.1 | - | 3 | 411 | 89 | ||
AK395083.1 | - | 4 | 484 | 89 | ||
AK398342.1 | - | 4 | 481 | 89 | ||
AK399143.1 | - | 4 | 494 | 89 | ||
AK400537.1 | - | 4 | 528 | 89 | ||
AY609757.1 | - | 4 | 477 | 89 | ||
D89492.1 | - | 2 | 152 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |