Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143928.1 | NP_001137400.1 | 2 | 5523 | 58 | |
GenBank mRNA | DQ006806.1 | - | 1 | 54 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |