Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214109.1 | NP_999274.1 | 7 | 746 | 65 | |
GenBank mRNA | D00237.1 | - | 745 | 65 | ||
M16651.1 | - | 7 | 746 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |