Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214237.2 | NP_999402.1 | 2 | 255 | 92 | |
XM_021068544.1 | XP_020924203.1 | 341 | 92 | |||
GenBank mRNA | AK344003.1 | - | 2 | 225 | 92 | |
EW238422.2 | - | 2 | 247 | 92 | ||
K01940.1 | - | 2 | 250 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |