Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214261.1 | NP_999426.1 | 2 | 1146 | 104 | |
XM_021078513.1 | XP_020934172.1 | 1371 | 104 | |||
XM_021078514.1 | XP_020934173.1 | 1308 | 104 | |||
XM_021078515.1 | XP_020934174.1 | 1503 | 104 | |||
XM_021078516.1 | XP_020934175.1 | 1589 | 104 | |||
XM_021078517.1 | XP_020934176.1 | 1271 | 104 | |||
GenBank mRNA | X04708.1 | - | 1146 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |