Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011727.1 | NP_001011727.1 | 5 - 6 | 655 | 60 | |
GenBank mRNA | AB527064.1 | - | 5 - 6 | 563 | 60 | |
AK234979.1 | - | 5 - 6 | 589 | 60 | ||
AK239495.1 | - | 5 - 6 | 689 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |