Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@1dd9a838
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Gene Expression
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss03393233_u1
See other SUMO2 GE Assays ›
Gene Symbol:
SUMO2
Gene Name:
small ubiquitin like modifier 2
Gene Aliases:
PSMT3, SMT3H2, Smt3A
Chromosome Location:
Chr.12: 6116694 - 6130089 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss03393233_u1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price (USD)
417.00
Your Price
Online offer (USD):
387.65
387.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
Displaying...
Build 4.1 Pig Chr.12: Ss03393233_u1
×
Created with Raphaël 2.1.2
SUMO2
NM_21398
4.1
AK240077
.1
AK237088
.1
6,037,00
0
6,038,00
0
6,039,00
0
6,040,00
0
6,041,00
0
6,042,00
0
6,043,00
0
6,044,00
0
6,045,00
0
6,046,00
0
6,047,00
0
6,048,00
0
6,049,00
0
6,050,00
0
6,051,00
0
6,052,00
0
6,053,00
0
6,054,00
0
6,055,00
0
6,056,00
0
6,057,00
0
6,058,00
0
6,059,00
0
6,060,00
0
6,061,00
0
6,062,00
0
6,063,00
0
6,064,00
0
6,065,00
0
6,066,00
0
6,067,00
0
6,068,00
0
6,069,00
0
6,070,00
0
6,071,00
0
6,072,00
0
6,073,00
0
6,074,00
0
6,075,00
0
6,076,00
0
6,077,00
0
6,078,00
0
6,079,00
0
6,080,00
0
6,081,00
0
6,082,00
0
6,083,00
0
6,084,00
0
6,085,00
0
6,086,00
0
6,087,00
0
6,088,00
0
6,089,00
0
6,090,00
0
6,091,00
0
6,092,00
0
6,093,00
0
6,094,00
0
6,095,00
0
6,096,00
0
6,097,00
0
6,098,00
0
6,099,00
0
6,100,00
0
6,101,00
0
6,102,00
0
6,103,00
0
6,104,00
0
6,105,00
0
6,106,00
0
6,107,00
0
6,108,00
0
6,109,00
0
6,110,00
0
6,111,00
0
6,112,00
0
6,113,00
0
6,114,00
0
6,115,00
0
6,116,00
0
6,117,00
0
6,118,00
0
6,119,00
0
6,120,00
0
6,121,00
0
6,122,00
0
6,123,00
0
6,124,00
0
6,125,00
0
6,126,00
0
6,127,00
0
6,128,00
0
6,129,00
0
6,130,00
0
6,131,00
0
6,132,00
0
6,133,00
0
6,134,00
0
Show all RefSeq and GenBank
Back To Top
Assay Details
Gene Symbol: SUMO2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_213984.1
NP_999149.1
4
640
128
GenBank mRNA
L77617.1
-
4
640
128
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Detect Genomic DNA
Recommended Assay
Sus scrofa
Both primers and probe map within a single exon
Gene Ontology Categories:
Function(s)
Process(es)
protein sumoylation
positive regulation of transcription from RNA polymerase II promoter
SUMO transferase activity
ubiquitin protein ligase binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-59d6677d6-m5kzq:80/100.66.73.217:80.
git-commit: 1e3318a2800c8d0a8829a508293490082e5b9833
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.30.0-Offline