Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129969.1 | NP_001123441.1 | 2 - 3 | 323 | 68 | |
GenBank mRNA | AF371322.1 | - | 2 - 3 | 323 | 68 | |
AK390435.1 | - | 2 - 3 | 469 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |