Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145222.2 | NP_001138694.1 | 1 - 2 | 124 | 70 | |
GenBank mRNA | AY609848.1 | - | 1 - 2 | 131 | 70 | |
EF208119.1 | - | 1 - 2 | 125 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |