Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190155.1 | NP_001177084.1 | 1 - 2 | 147 | 90 | |
XM_021071897.1 | XP_020927556.1 | 168 | 90 | |||
GenBank mRNA | AK236996.1 | - | 1 - 2 | 146 | 90 | |
AK390036.1 | - | 1 - 2 | 146 | 90 | ||
AY610358.1 | - | 1 - 2 | 128 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |