Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243406.1 | NP_001230335.1 | 2 - 3 | 651 | 98 | |
GenBank mRNA | AK350830.1 | - | 2 - 3 | 653 | 98 | |
AK397123.1 | - | 2 - 3 | 682 | 98 | ||
FD629736.1 | - | 2 - 3 | 235 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |