Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244809.1 | NP_001231738.1 | 3 - 4 | 364 | 67 | |
GenBank mRNA | AK344004.1 | - | 3 - 4 | 365 | 67 | |
AK400047.1 | - | 3 - 4 | 376 | 67 | ||
AY610478.1 | - | 3 - 4 | 297 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |