Save up to 49% with protein expression workflow promo

Save now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Ss06886154_g1
            See other LAMTOR3 GE Assays ›
            Gene Symbol:
            LAMTOR3
            Gene Name:
            late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
            Gene Aliases:
            -
            Chromosome Location:
            Chr.8: 120625897 - 120642630 on Build Sscrofa11.1
            Species:
            Pig
            Species Specific ID (Flybase ID):
            -
            Assay ID Ss06886154_g1
            Best Coverage
            Size
            Availability Made To Order
            Catalog # 4351372
            Price
            Your Price
            Online offer:
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data.
            ×
            Back To Top

            Assay Details

            Gene Symbol: LAMTOR3
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq XM_003129301.4 XP_003129349.1 490 76
            XM_021101726.1 XP_020957385.1 3528 76
            GenBank mRNA AK231033.1 - 6 - 7 439 76
            AK393794.1 - 6 - 7 478 76
            AY610499.1 - 6 - 7 358 76

            Back To Top

            More Information


            Set Membership:

            Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

            Panther Classification:

            Molecular Function -

            scaffold/adaptor protein

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-wfwm6:80/100.66.76.90:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0