Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002270372.1 | XP_002270408.1 | 3 - 4 | 362 | 67 | |
GenBank mRNA | DQ504308.1 | - | 3 - 4 | 368 | 67 | |
DQ871590.1 | - | 3 - 4 | 333 | 67 | ||
EF157728.1 | - | 3 - 4 | 341 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Vitis vinifera Probe spans exons |