Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004781.1 | NP_001004781.1 | 4 - 5 | 456 | 85 | |
GenBank mRNA | BC074516.1 | - | 4 - 5 | 456 | 85 | |
CR926214.2 | - | 4 - 5 | 360 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |