Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203521.1 | NP_988852.1 | 1 - 2 | 127 | 92 | |
GenBank mRNA | AY079194.1 | - | 1 - 2 | 127 | 92 | |
CR762022.2 | - | 1 - 2 | 99 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |