Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203809.1 | NP_989140.1 | 3 - 4 | 453 | 94 | |
XM_012968153.2 | XP_012823607.1 | 3 - 4 | 313 | 94 | ||
GenBank mRNA | BC061401.1 | - | 3 - 4 | 453 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |