Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005122.2 | NP_001005122.1 | 3 - 4 | 260 | 72 | |
GenBank mRNA | BC077657.1 | - | 3 - 4 | 261 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |