Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005455.1 | NP_001005455.1 | 6 - 7 | 796 | 98 | |
GenBank mRNA | BC074571.1 | - | 6 - 7 | 796 | 98 | |
CR855832.2 | - | 6 - 7 | 742 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |