Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005643.1 | NP_001005643.1 | 2 - 3 | 469 | 67 | |
XM_012954970.2 | XP_012810424.1 | 2 - 3 | 229 | 67 | ||
XM_012954971.2 | XP_012810425.1 | 2 - 3 | 225 | 67 | ||
XM_012954972.2 | XP_012810426.1 | 2 - 3 | 198 | 67 | ||
GenBank mRNA | BC074643.1 | - | 2 - 3 | 469 | 67 | |
CR760378.2 | - | 1 - 2 | 134 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |