Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005679.1 | NP_001005679.1 | 3 - 4 | 497 | 97 | |
XM_012958815.2 | XP_012814269.2 | 4 - 5 | 391 | 97 | ||
GenBank mRNA | BC075099.1 | - | 3 - 4 | 497 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |