Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pla2g12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007916.1 | NP_001007917.1 | 2 - 3 | 322 | 69 | |
GenBank mRNA | BC080358.1 | - | 2 - 3 | 322 | 69 | |
CR926443.2 | - | 2 - 3 | 368 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |