Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008181.1 | NP_001008182.2 | 1039 | 64 | ||
GenBank mRNA | BC082505.1 | - | 1039 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis |