Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008197.2 | NP_001008198.1 | 14 - 15 | 1272 | 104 | |
XM_012970351.1 | XP_012825805.1 | 14 - 15 | 1811 | 104 | ||
XM_018097092.1 | XP_017952581.1 | 14 - 15 | 1264 | 104 | ||
GenBank mRNA | BC082526.1 | - | 14 - 15 | 1273 | 104 | |
CR760737.2 | - | 14 - 15 | 1255 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |