Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011190.1 | NP_001011190.1 | 1 - 2 | 340 | 65 | |
GenBank mRNA | BC086506.2 | - | 1 - 2 | 340 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |