Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cxorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204012.2 | NP_989343.1 | 5 - 6 | 540 | 96 | |
XM_018096190.1 | XP_017951679.1 | 5 - 6 | 596 | 96 | ||
GenBank mRNA | BC064195.1 | - | 5 - 6 | 521 | 96 | |
BC084906.1 | - | 5 - 6 | 540 | 96 | ||
CR760470.2 | - | 5 - 6 | 521 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |