Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult1c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011489.1 | NP_001011489.1 | 5 - 6 | 588 | 91 | |
XM_012954435.1 | XP_012809889.1 | 11 - 12 | 1369 | 91 | ||
GenBank mRNA | BC088870.1 | - | 5 - 6 | 588 | 91 |
Gene Symbol: LOC100487424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002941304.4 | XP_002941350.1 | 5 - 6 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |