Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016671.2 | NP_001016671.1 | 5 - 6 | 722 | 62 | |
GenBank mRNA | BC157339.1 | - | 5 - 6 | 512 | 62 | |
BC170616.1 | - | 5 - 6 | 693 | 62 | ||
BC170618.1 | - | 5 - 6 | 693 | 62 | ||
CR760242.2 | - | 5 - 6 | 722 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |