Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016521.2 | NP_001016521.1 | 807 | 110 | ||
GenBank mRNA | BC170807.1 | - | 6 - 7 | 741 | 110 | |
BC170809.1 | - | 6 - 7 | 741 | 110 | ||
CR926207.2 | - | 807 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |