Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017032.2 | NP_001017032.1 | 3 - 4 | 390 | 97 | |
GenBank mRNA | BC135727.1 | - | 3 - 4 | 356 | 97 | |
BC157282.1 | - | 3 - 4 | 410 | 97 | ||
CR762013.2 | - | 3 - 4 | 390 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |