Search Thermo Fisher Scientific
Gene Symbol: cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017228.3 | NP_001017228.1 | 5 - 6 | 486 | 71 | |
GenBank mRNA | BC158921.1 | - | 1 - 2 | 59 | 71 | |
BC167872.1 | - | 2 - 3 | 212 | 71 | ||
BC168096.1 | - | 2 - 3 | 224 | 71 | ||
CR760272.2 | - | 5 - 6 | 471 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |