Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017237.2 | NP_001017237.1 | 1 - 2 | 137 | 79 | |
GenBank mRNA | BC157296.1 | - | 1 - 2 | 123 | 79 | |
BC157341.1 | - | 1 - 2 | 123 | 79 | ||
CR760203.2 | - | 1 - 2 | 137 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |