Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203964.3 | NP_989295.2 | 4 - 5 | 548 | 99 | |
GenBank mRNA | BC064234.1 | - | 4 - 5 | 455 | 99 | |
CN080459.1 | - | 4 - 5 | 485 | 99 | ||
CR926434.1 | - | 4 - 5 | 529 | 99 | ||
CX876971.1 | - | 4 - 5 | 548 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |