Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213670.2 | NP_998835.1 | 4 - 5 | 694 | 78 | |
GenBank mRNA | BC067911.1 | - | 4 - 5 | 581 | 78 | |
CR761289.1 | - | 4 - 5 | 694 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |