Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045668.1 | NP_001039133.1 | 1 - 2 | 304 | 74 | |
XM_012968554.2 | XP_012824008.1 | 1 - 2 | 1225 | 74 | ||
GenBank mRNA | BC160584.1 | - | 1 - 2 | 273 | 74 | |
CR942498.2 | - | 1 - 2 | 304 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |