Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078817.1 | NP_001072285.1 | 12 - 13 | 2645 | 65 | |
XM_012952548.2 | XP_012808002.1 | 12 - 13 | 3199 | 65 | ||
GenBank mRNA | BC121308.1 | - | 12 - 13 | 2645 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |