Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tesc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078955.1 | NP_001072423.1 | 3 - 4 | 532 | 82 | |
GenBank mRNA | BC121662.1 | - | 3 - 4 | 532 | 82 | |
BC135886.1 | - | 3 - 4 | 384 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |