Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: c1orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079184.1 | NP_001072652.1 | 2 - 3 | 281 | 82 | |
XM_012961365.2 | XP_012816819.1 | 2 - 3 | 313 | 82 | ||
XM_018093103.1 | XP_017948592.1 | 2 - 3 | 301 | 82 | ||
GenBank mRNA | BC123935.1 | - | 2 - 3 | 281 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |