Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079247.1 | NP_001072715.1 | 6 - 7 | 662 | 71 | |
XM_012961367.2 | XP_012816821.1 | 6 - 7 | 870 | 71 | ||
GenBank mRNA | BC125674.1 | - | 6 - 7 | 662 | 71 | |
CT025320.2 | - | 6 - 7 | 660 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |